Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-5-methyl-N-(quinolin-5-yl)indoline-1-carboxamide 98%

Supplier: AMBEED, INC
Warning

A973943-10MG A973943-1MG
76656-422EA 85 USD InStock
76656-422 76656-424
6-Chloro-5-methyl-N-(quinolin-5-yl)indoline-1-carboxamide 98%
6-Chloro-5-methyl-N-(quinolin-5-yl)indoline-1-carboxamide
Precaución: for Research Use Only


Formula: C₁₉H₁₆ClN₃O
MDL Number: MFCD05664741
CAS Number: 162100-15-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty