Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trans-4-(benzyloxy)cyclohexane-1-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A979731-100MG A979731-1G A979731-250MG
76656-512EA 71 USD InStock
76656-512 76656-514 76656-516
Trans-4-(benzyloxy)cyclohexane-1-carboxylic acid 97%
Trans-4-(benzyloxy)cyclohexane-1-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₈O₃
MDL Number: MFCD31640824
CAS Number: 1558003-77-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty