Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,3,3-Tetramethyl-4,7,10,13-tetraoxa-3-silapentadecan-15-ol 95%

Supplier: AMBEED, INC
Danger

A984366-1G A984366-250MG A984366-5G A984366-25G
76656-826EA 27 USD InStock
76656-826 76656-828 76656-830 77623-272
2,2,3,3-Tetramethyl-4,7,10,13-tetraoxa-3-silapentadecan-15-ol 95%
2,2,3,3-Tetramethyl-4,7,10,13-tetraoxa-3-silapentadecan-15-ol
Precaución: for Research Use Only


Formula: C₁₄H₃₂O₅Si
MDL Number: MFCD25424114
CAS Number: 134179-40-1
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty