Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-6,7-dihydro-5h-pyrazolo[5,1-b][1,3]oxazine 95%

Supplier: AMBEED, INC
Warning

A987735-100MG A987735-250MG A987735-50MG
76657-130EA 177 USD InStock
76657-130 76657-132 76657-134
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-6,7-dihydro-5h-pyrazolo[5,1-b][1,3]oxazine 95%
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-6,7-dihydro-5h-pyrazolo[5,1-b][1,3]oxazine
Precaución: for Research Use Only


Formula: C₁₂H₁₉BN₂O₃
MDL Number: MFCD22415472
CAS Number: 1383675-83-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty