Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N1,N1-Bis(pyridin-2-ylmethyl)ethane-1,2-diamine 97%

Supplier: AMBEED, INC
Warning

A1351750-100MG A1351750-250MG
76658-226EA 133 USD InStock
76658-226 76658-228
N1,N1-Bis(pyridin-2-ylmethyl)ethane-1,2-diamine 97%
N1,N1-Bis(pyridin-2-ylmethyl)ethane-1,2-diamine
Precaución: for Research Use Only


Formula: C₁₄H₁₈N₄
MDL Number: MFCD20527984
CAS Number: 189440-33-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty