Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2'-(Trifluoromethyl)propiophenone 97%

Supplier: AMBEED, INC
Warning

A1368204-10G A1368204-25G
76659-460EA 35 USD InStock
76659-460 76659-462
2'-(Trifluoromethyl)propiophenone 97%
2'-(Trifluoromethyl)propiophenone
Precaución: for Research Use Only


Formula: C₁₀H₉F₃O
MDL Number: MFCD00013557
CAS Number: 16185-96-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty