Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxypropyl 3,4-difluorobenzoate 98%

Supplier: AMBEED, INC
Warning

A1417630-1G A1417630-250MG A1417630-5G
76660-672EA 25 USD InStock
76660-672 76660-674 76660-676
3-Hydroxypropyl 3,4-difluorobenzoate 98%
3-Hydroxypropyl 3,4-difluorobenzoate
Precaución: for Research Use Only


Formula: C₁₀H₁₀F₂O₃
CAS Number: 684648-49-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty