Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(1-Methyl-1H-imidazol-3-ium-3-yl)propane-1-sulfonate 98%

Supplier: AMBEED, INC
Warning

A1420893-25G A1420893-5G
76660-700EA 35 USD InStock
76660-700 76660-702
3-(1-Methyl-1H-imidazol-3-ium-3-yl)propane-1-sulfonate 98%
3-(1-Methyl-1H-imidazol-3-ium-3-yl)propane-1-sulfonate
Precaución: for Research Use Only


Formula: C₇H₁₂N₂O₃S
MDL Number: MFCD30497948
CAS Number: 148019-49-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty