Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Diisopropyl-1H-benzo[d]imidazol-3-ium bromide 98%

Supplier: AMBEED, INC
Warning

A1442325-1G A1442325-250MG A1442325-25G A1442325-5G
76661-056EA 118 USD InStock
76661-056 76661-058 77622-096 77622-098
1,3-Diisopropyl-1H-benzo[d]imidazol-3-ium bromide 98%
1,3-Diisopropyl-1H-benzo[d]imidazol-3-ium bromide
Precaución: for Research Use Only


Formula: C₁₃H₁₉BrN₂
MDL Number: MFCD33398687
CAS Number: 684283-51-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty