Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-(Pyridin-2-yl)piperidin-4-yl)methanamine 97%

Supplier: AMBEED, INC
Danger

A1452173-100MG A1452173-1G A1452173-250MG A1452173-50MG
76661-434EA 213 USD InStock
76661-434 76661-436 76661-438 76661-440
(4-(Pyridin-2-yl)piperidin-4-yl)methanamine 97%
(4-(Pyridin-2-yl)piperidin-4-yl)methanamine
Precaución: for Research Use Only


Formula: C₁₁H₁₇N₃
MDL Number: MFCD23924134
CAS Number: 1501149-78-5
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty