Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyldimethylstearylammonium chloride 98% + (mixture of isomers) (contains< 5% H₂O)

Supplier: AMBEED, INC
Danger

A1452303-25G A1452303-5G
76661-458EA 19 USD InStock
76661-458 76661-460
Benzyldimethylstearylammonium chloride 98% + (mixture of isomers) (contains< 5% H₂O)
Benzyldimethylstearylammonium chloride
Precaución: for Research Use Only


Formula: C₂₇H₅₀ClN
MDL Number: MFCD00011776
CAS Number: 122-19-0
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty