Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methoxy-3,4-dihydronaphthalen-1-yl trifluoromethanesulfonate 95%

Supplier: AMBEED, INC
Danger

A1452518-100MG A1452518-250MG
76661-510EA 31 USD InStock
76661-510 76661-512
6-Methoxy-3,4-dihydronaphthalen-1-yl trifluoromethanesulfonate 95%
6-Methoxy-3,4-dihydronaphthalen-1-yl trifluoromethanesulfonate
Precaución: for Research Use Only


Formula: C₁₂H₁₁F₃O₄S
CAS Number: 115375-59-2
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty