3,4-Difluorotoluene 99%

Supplier: AMBEED, INC
Danger

A1475866-1G A1475866-5G
76663-442EA 6 USD
76663-442 76663-444
3,4-Difluorotoluene 99%
3,4-Difluorotoluene

Precaución: for Research Use Only

Formula: C₇H₆F₂
MW: 128.12
Density: 1.12
MDL Number: MFCD00075087
CAS Number: 2927-34-6
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR