Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(tert-Butyl) 4-ethyl 3-oxo-2,3-dihydro-1H-pyrazole-1,4-dicarboxylate 98%

Supplier: AMBEED, INC
Warning

A1476041-100G A1476041-25G A1476041-5G
76663-604EA 50 USD InStock
76663-604 76663-606 76663-608
1-(tert-Butyl) 4-ethyl 3-oxo-2,3-dihydro-1H-pyrazole-1,4-dicarboxylate 98%
1-(tert-Butyl) 4-ethyl 3-oxo-2,3-dihydro-1H-pyrazole-1,4-dicarboxylate
Precaución: for Research Use Only


Formula: C₁₁H₁₆N₂O₅
MDL Number: MFCD13689095
CAS Number: 178424-17-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty