Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-Bromophenyl)-N-methylbutyramide 97%

Supplier: AMBEED, INC
Warning

A1485929-100MG A1485929-1G A1485929-250MG A1485929-5G
76665-020EA 21 USD InStock
76665-020 76665-022 76665-024 76665-026
N-(4-Bromophenyl)-N-methylbutyramide 97%
N-(4-Bromophenyl)-N-methylbutyramide
Precaución: for Research Use Only


Formula: C₁₁H₁₄BrNO
CAS Number: 911230-59-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty