Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[1,1':4',1'':4'',1'''-Quaterphenyl]-3,3''',5,5'''-tetracarbaldehyde 97%

Supplier: AMBEED, INC
Warning

A1489886-100MG A1489886-1G A1489886-250MG
76665-548EA 24 USD InStock
76665-548 76665-550 76665-552
[1,1':4',1'':4'',1'''-Quaterphenyl]-3,3''',5,5'''-tetracarbaldehyde 97%
[1,1':4',1'':4'',1'''-Quaterphenyl]-3,3''',5,5'''-tetracarbaldehyde
Precaución: for Research Use Only


Formula: C₂₈H₁₈O₄
MDL Number: MFCD34167549
CAS Number: 2459652-59-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty