Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dioxoisoindolin-2-yl 2-ethylbutanoate 97%

Supplier: AMBEED, INC
Warning

A1496198-1G A1496198-250MG A1496198-5G
76666-504EA 58 USD InStock
76666-504 76666-506 76666-508
1,3-Dioxoisoindolin-2-yl 2-ethylbutanoate 97%
1,3-Dioxoisoindolin-2-yl 2-ethylbutanoate
Precaución: for Research Use Only


Formula: C₁₄H₁₅NO₄
CAS Number: 2245137-17-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty