Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N1,N1,N2,N2-Tetramethyloxalamide 95%

Supplier: AMBEED, INC
Danger

A1516734-1G A1516734-5G
76667-506EA 31 USD InStock
76667-506 76667-508
N1,N1,N2,N2-Tetramethyloxalamide 95%
N1,N1,N2,N2-Tetramethyloxalamide
Precaución: for Research Use Only


Formula: C₆H₁₂N₂O₂
MDL Number: MFCD00025670
CAS Number: 1608-14-6
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty