Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Borono-3-(trifluoromethyl)benzoic acid 95%

Supplier: AMBEED, INC
Warning

A1532111-100MG A1532111-1G A1532111-250MG
76667-958EA 39 USD InStock
76667-958 76667-960 76667-962
4-Borono-3-(trifluoromethyl)benzoic acid 95%
4-Borono-3-(trifluoromethyl)benzoic acid
Precaución: for Research Use Only


Formula: C₈H₆BF₃O₄
MDL Number: MFCD23380382
CAS Number: 1383969-65-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty