Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl N-(1,2,3,4-tetrahydroisoquinolin-5-yl)carbamate hydrochloride 95%

Supplier: AMBEED, INC
Warning

A1015151-250MG A1015151-50MG
76671-102EA 47 USD InStock
76671-102 76671-104
Methyl N-(1,2,3,4-tetrahydroisoquinolin-5-yl)carbamate hydrochloride 95%
Methyl N-(1,2,3,4-tetrahydroisoquinolin-5-yl)carbamate hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₁₅ClN₂O₂
MDL Number: MFCD22378756
CAS Number: 1394041-21-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty