Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Morpholin-4-yl)propane-1-sulfonyl chloride hydrochloride 98%

Supplier: AMBEED, INC
Danger

A1032273-1G A1032273-250MG
76671-260EA 231 USD InStock
76671-260 76671-262
3-(Morpholin-4-yl)propane-1-sulfonyl chloride hydrochloride 98%
3-(Morpholin-4-yl)propane-1-sulfonyl chloride hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₅Cl₂NO₃S
MDL Number: MFCD18425900
CAS Number: 197891-91-1
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty