Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyanomethyl Methyl(phenyl)carbamodithioate 95%

Supplier: AMBEED, INC
Warning

A1130894-1G A1130894-250MG
76672-736EA 88 USD InStock
76672-736 76672-738
Cyanomethyl Methyl(phenyl)carbamodithioate 95%
Cyanomethyl Methyl(phenyl)carbamodithioate
Precaución: for Research Use Only


Formula: C₁₀H₁₀N₂S₂
MDL Number: MFCD16038113
CAS Number: 76926-16-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty