Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 5-hydroxypentanoate 95%

Supplier: AMBEED, INC
Warning

A1145668-100G A1145668-10G A1145668-1G A1145668-5G
76673-328EA 396 USD InStock
76673-328 76673-330 76673-332 76673-334
Sodium 5-hydroxypentanoate 95%
Sodium 5-hydroxypentanoate
Precaución: for Research Use Only


Formula: C₅H₉O₃·Na
MW: 140.113 g/mol
MDL Number: MFCD22192345
CAS Number: 37435-69-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty