Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

AUDA 98%

Supplier: AMBEED, INC
Warning

A1148766-10MG A1148766-1MG A1148766-25MG A1148766-50MG A1148766-5MG
76673-794EA 48 USD InStock
76673-794 76673-796 76673-798 76673-800 76673-802
AUDA 98%
AUDA
Precaución: for Research Use Only


Formula: C₂₁H₃₆N₂O₃
MDL Number: MFCD12912267
CAS Number: 479413-70-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty