Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-((3,5-Dichlorophenyl)sulfonyl)-N-(4-fluorobenzyl)piperidine-4-carboxamide 99%

Supplier: AMBEED, INC
Warning

A1149110-10MG A1149110-1MG A1149110-25MG A1149110-5MG
76673-854EA 42 USD InStock
76673-854 76673-856 76673-858 76673-860
1-((3,5-Dichlorophenyl)sulfonyl)-N-(4-fluorobenzyl)piperidine-4-carboxamide 99%
1-((3,5-Dichlorophenyl)sulfonyl)-N-(4-fluorobenzyl)piperidine-4-carboxamide
Precaución: for Research Use Only


Formula: C₁₉H₁₉Cl₂FN₂O₃S
MDL Number: MFCD31916240
CAS Number: 2227549-00-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty