Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-1,3,4-trimethylbenzene 98%

Supplier: AMBEED, INC
Warning

A1150274-25G A1150274-5G
76674-212EA 64 USD InStock
76674-212 76674-214
2-Methoxy-1,3,4-trimethylbenzene 98%
2-Methoxy-1,3,4-trimethylbenzene
Precaución: for Research Use Only


Formula: C₁₀H₁₄O
MW: 150.2176 g/mol
MDL Number: MFCD28362204
CAS Number: 21573-36-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty