Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,5'-(1,3,6,8-Tetraoxobenzo[lmn][3,8]phenanthroline-2,7(1H,3H,6H,8H)-diyl)diisophthalic acid 98%

Supplier: AMBEED, INC
Warning

A1156784-10G A1156784-250MG A1156784-5G
76675-100EA 296 USD InStock
76675-100 76675-102 76675-104
5,5'-(1,3,6,8-Tetraoxobenzo[lmn][3,8]phenanthroline-2,7(1H,3H,6H,8H)-diyl)diisophthalic acid 98%
5,5'-(1,3,6,8-Tetraoxobenzo[lmn][3,8]phenanthroline-2,7(1H,3H,6H,8H)-diyl)diisophthalic acid
Precaución: for Research Use Only


Formula: C₃₀H₁₄N₂O₁₂
MDL Number: MFCD00306176
CAS Number: 350024-36-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty