Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-(Benzo[c][1,2,5]thiadiazole-4,7-diyl)dibenzaldehyde 98%

Supplier: AMBEED, INC
Warning

A1195794-100MG A1195794-1G A1195794-250MG
76677-348EA 9 USD InStock
76677-348 76677-350 76677-352
4,4'-(Benzo[c][1,2,5]thiadiazole-4,7-diyl)dibenzaldehyde 98%
4,4'-(Benzo[c][1,2,5]thiadiazole-4,7-diyl)dibenzaldehyde
Precaución: for Research Use Only


Formula: C₂₀H₁₂N₂O₂S
CAS Number: 914651-17-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty