Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(3-((4aS,7aS)-2-Amino-4a,5,7,7a-tetrahydro-4H-furo[3,4-d][1,3]thiazin-7a-yl)-4-fluorophenyl)-5-fluoropicolinamide hydrochloride 98%

Supplier: AMBEED, INC
Warning

A1226355-10MG A1226355-1MG A1226355-25MG A1226355-50MG A1226355-5MG
76679-374EA 64 USD InStock
76679-374 76679-376 76679-378 76679-380 76679-382
N-(3-((4aS,7aS)-2-Amino-4a,5,7,7a-tetrahydro-4H-furo[3,4-d][1,3]thiazin-7a-yl)-4-fluorophenyl)-5-fluoropicolinamide hydrochloride 98%
N-(3-((4aS,7aS)-2-Amino-4a,5,7,7a-tetrahydro-4H-furo[3,4-d][1,3]thiazin-7a-yl)-4-fluorophenyl)-5-fluoropicolinamide hydrochloride
Precaución: for Research Use Only


Formula: C₁₈H₁₇ClF₂N₄O₂S
MDL Number: MFCD24387533
CAS Number: 1262036-49-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty