Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2,2-Dimethoxyethyl)-1H-pyrazole 97%

Supplier: AMBEED, INC
Warning

A1228673-100MG A1228673-1G A1228673-250MG
76679-676EA 13 USD InStock
76679-676 76679-678 76679-680
1-(2,2-Dimethoxyethyl)-1H-pyrazole 97%
1-(2,2-Dimethoxyethyl)-1H-pyrazole
Precaución: for Research Use Only


Formula: C₇H₁₂N₂O₂
MDL Number: MFCD21728287
CAS Number: 876164-61-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty