Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1-(1-Isoquinolinyl)-2-naphthyl)diphenylphosphine 97%

Supplier: AMBEED, INC
Warning

A1258466-100MG A1258466-1G A1258466-250MG A1258466-5G
76680-762EA 101 USD InStock
76680-762 76680-764 76680-766 77621-352
(1-(1-Isoquinolinyl)-2-naphthyl)diphenylphosphine 97%
(1-(1-Isoquinolinyl)-2-naphthyl)diphenylphosphine
Precaución: for Research Use Only


Formula: C₃₁H₂₂NP
MDL Number: MFCD01073773
CAS Number: 149245-03-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty