Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(tert-Butoxycarbonyl)-L-leucine hydrate 97% (contains < 10% H₂O)

Supplier: AMBEED, INC
Warning

A1270494-100G A1270494-25G A1270494-500G A1270494-1000G
76681-048EA 18 USD InStock
76681-048 76681-050 76681-052 77621-396
N-(tert-Butoxycarbonyl)-L-leucine hydrate 97% (contains < 10% H₂O)
N-(tert-Butoxycarbonyl)-L-leucine hydrate
Precaución: for Research Use Only


Formula: C₁₁H₂₁NO₄·xH₂O
MDL Number: MFCD00066067
CAS Number: 13139-15-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty