Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-butyl 3-oxo-2,3-Dhydro-1H-pyrazole-1-carboxylate 98%

Supplier: AMBEED, INC
Warning

A1276709-250MG A1276709-5G A1276709-1G A1276709-10G
76681-450EA 55 USD InStock
76681-450 76681-452 77596-676 77621-446
tert-butyl 3-oxo-2,3-Dhydro-1H-pyrazole-1-carboxylate 98%
tert-butyl 3-oxo-2,3-Dhydro-1H-pyrazole-1-carboxylate
Precaución: for Research Use Only


Formula: C₈H₁₂N₂O₃
MDL Number: MFCD31742743
CAS Number: 2088458-39-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty