Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Di(pyridin-3-yl)-1,3,4-thiadiazole 97%

Supplier: AMBEED, INC
Warning

A1351734-1G A1351734-250MG A1351734-5G
76682-100EA 167 USD InStock
76682-100 76682-102 76682-104
2,5-Di(pyridin-3-yl)-1,3,4-thiadiazole 97%
2,5-Di(pyridin-3-yl)-1,3,4-thiadiazole
Precaución: for Research Use Only


Formula: C₁₂H₈N₄S
MDL Number: MFCD18430981
CAS Number: 15362-52-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty