Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Azelnidipine 97%

Supplier: AMBEED, INC
Warning

A110442-100MG A110442-25MG A110442-50MG
76687-510EA 9 USD InStock
76687-510 76687-512 76687-514
Azelnidipine 97%
Azelnidipine
Precaución: for Research Use Only


Formula: C₃₃H₃₄N₄O₆
MDL Number: MFCD00865803
CAS Number: 123524-52-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty