Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Deracoxib 98%

Supplier: AMBEED, INC
Warning

A116985-10MG A116985-50MG A116985-100MG A116985-1G A116985-1MG A116985-250MG
76687-914EA 6 USD InStock
76687-914 76687-916 77623-978 77623-980 77623-982 77623-984
Deracoxib 98%
Deracoxib
Precaución: for Research Use Only


Formula: C₁₇H₁₄ClF₃N₃O₃S
MDL Number: MFCD09837763
CAS Number: 169590-41-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty