Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-2-Abz-OH 95%

Supplier: AMBEED, INC
Warning

A156016-100G A156016-10G A156016-1G A156016-25G A156016-5G
76689-558EA 279 USD InStock
76689-558 76689-560 76689-562 76689-564 76689-566
Boc-2-Abz-OH 95%
Boc-2-Abz-OH
Precaución: for Research Use Only


Formula: C₁₂H₁₅NO₄
MDL Number: MFCD00151862
CAS Number: 68790-38-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty