Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Risocaine (propyl-4-aminobenzoate) 98%

Supplier: AMBEED, INC
Warning

A160089-100G A160089-25G A160089-5G
76689-790EA 224 USD InStock
76689-790 76689-792 76689-794
Risocaine (propyl-4-aminobenzoate) 98%
Risocaine (propyl-4-aminobenzoate)
Precaución: for Research Use Only


Formula: C₁₀H₁₃NO₂
MDL Number: MFCD00017111
CAS Number: 94-12-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty