Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9H-Carbazole-3-carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A165674-100MG A165674-1G A165674-250MG
76690-020EA 21 USD InStock
76690-020 76690-022 76690-024
9H-Carbazole-3-carboxylic acid 95%
9H-Carbazole-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₃H₉NO₂
MDL Number: MFCD03152409
CAS Number: 51035-17-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty