1-(2-Methylimidazo[1,2-a]pyridin-3-yl)ethanone 95%

Supplier: AMBEED, INC
Warning

A246331-250MG A246331-5G
76692-364EA 25 USD
76692-364 76692-366
1-(2-Methylimidazo[1,2-a]pyridin-3-yl)ethanone 95%
1-(2-Methylimidazo[1,2-a]pyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O
MW: 174.2000 g/mol
MDL Number: MFCD00665892
CAS Number: 29096-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR