Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzylidenebis(tricyclohexylphosphine)dichlororuthenium 98%

Supplier: AMBEED, INC
Danger

A260995-250MG A260995-1G
76692-706EA 8 USD InStock
76692-706 77550-642
Benzylidenebis(tricyclohexylphosphine)dichlororuthenium 98%
Benzylidenebis(tricyclohexylphosphine)dichlororuthenium
Precaución: For research use only.


Formula: C₄₃H₇₂Cl₂P₂Ru
MW: 822.9553 g/mol
MDL Number: MFCD01090946
CAS Number: 172222-30-9
UN: 1325
ADR: 4.1 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty