Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxyindole 98%

Supplier: AMBEED, INC
Warning

A310676-1G A310676-250MG A310676-25G A310676-5G
76693-572EA 11 USD InStock
76693-572 76693-574 76693-576 76693-578
4-Hydroxyindole 98%
4-Hydroxyindole
Precaución: for Research Use Only


Formula: C₈H₇NO
MDL Number: MFCD00005667
CAS Number: 2380-94-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty