Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethynyl-N,N-dimethylaniline 96%

Supplier: AMBEED, INC
Warning

A339877-250MG A339877-25G A339877-5G
76693-952EA 27 USD InStock
76693-952 76693-954 77550-976
4-Ethynyl-N,N-dimethylaniline 96%
4-Ethynyl-N,N-dimethylaniline
Precaución: for Research Use Only


Formula: C₁₀H₁₁N
MDL Number: MFCD00168816
CAS Number: 17573-94-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty