Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(Benzoyloxy)succinimide 97%

Supplier: AMBEED, INC
Danger

A381164-1G A381164-250MG A381164-25G
76694-474EA 24 USD InStock
76694-474 76694-476 76694-478
N-(Benzoyloxy)succinimide 97%
N-(Benzoyloxy)succinimide
Precaución: for Research Use Only


Formula: C₁₁H₉NO₄
MW: 219.196 g/mol
MDL Number: MFCD00078953
CAS Number: 23405-15-4
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty