6-tert-Butyldimethylsilyloxy-2-naphthaleneboronic acid 98%

Supplier: AMBEED, INC
Warning

A393687-1G A393687-250MG A393687-5G
76694-708EA 81 USD
76694-708 76694-710 76694-712
6-tert-Butyldimethylsilyloxy-2-naphthaleneboronic acid 98%
6-tert-Butyldimethylsilyloxy-2-naphthaleneboronic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₃BO₃Si
MW: 302.2485 g/mol
MDL Number: MFCD04115644
CAS Number: 179942-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR