Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-Asp(OtBu)-ONp 95%

Supplier: AMBEED, INC
Warning

A539163-10G A539163-1G A539163-5G
76696-018EA 69 USD InStock
76696-018 76696-020 76696-022 76696-024
Boc-Asp(OtBu)-ONp 95%
Boc-Asp(OtBu)-ONp
Precaución: for Research Use Only


Formula: C₁₉H₂₆N₂O₈
MDL Number: MFCD00076913
CAS Number: 29365-05-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty