Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Decyltrimethylammonium bromide 97%

Supplier: AMBEED, INC
Warning

A569548-100G A569548-10G A569548-25G A569548-5G A569548-500G
76696-284EA 65 USD InStock
76696-284 76696-286 76696-288 76696-290 77627-180
Decyltrimethylammonium bromide 97%
Decyltrimethylammonium bromide
Precaución: for Research Use Only


Formula: C₁₃H₃₀BrN
MDL Number: MFCD00041973
CAS Number: 2082-84-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty