Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(cis-Tetrahydrofuran-2,5-diyl)dimethanol 80%

Supplier: AMBEED, INC
Warning

A609454-100MG A609454-1G A609454-250MG A609454-25G A609454-5G
76696-600EA 45 USD InStock
76696-600 76696-602 76696-604 76696-606 76696-608
(cis-Tetrahydrofuran-2,5-diyl)dimethanol 80%
(cis-Tetrahydrofuran-2,5-diyl)dimethanol
Precaución: for Research Use Only


Formula: C₆H₁₂O₃
MDL Number: MFCD07369849
CAS Number: 2144-40-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty