Phenol red 95%

Supplier: AMBEED, INC
Warning

A122951-100G A122951-10G A122951-5G
76718-632EA 51 USD
76718-632 76718-634 76718-636
Phenol red 95%
Phenol red

Precaución: for Research Use Only

Formula: C₁₉H₁₄O₅S
MW: 354.38 g/mol
MDL Number: MFCD00003552
CAS Number: 143-74-8
Merck Index: 13,07329

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR