Pyridine iodine monochloride 98%
Supplier: AMBEED, INC
|
Warning
|
A155009-100MG
A155009-1G
A155009-250MG
A155009-5G
76719-910EA
17
USD
76719-910
76719-912
76719-914
77667-196
Pyridine iodine monochloride 98%
Pyridine iodine monochloride
Precaución: For research use only.
Formula:
C₅H₅N MW: 241.46 g/mol |
MDL Number:
MFCD06410987 CAS Number: 6443-90-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...